Please login to your online account to display your discounted pricing

Fisherbrand™ Automatic Burets with Stopcock


Ideal for storing standard solutions

Manufacturer: fisherbrand™  720823

 View more versions of this product

Catalog No. 03-842-2B

  • / Each
 

Description & Specifications

Specifications

Color Amber
Includes Amber glass reservoir bottle, rubber stopper, one glass buret with central filling tube, rubber bulb, glass and rubber tubing connections, and glass stopcock
Material Glass
Product Type Automatic Buret
For Use With (Application) Ideal for storing standard solutions
Subdivision 0.1mL
Capacity (Metric) 25mL