missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Automatic Burets with Pinch Valve

With storage bottle

$198.07 - $199.50

Specifications

Capacity (Metric) 25mL
Product Type Automatic Buret
Subdivision 0.1mL
Material Glass
View More Specs

Includes: Glass buret with central filling tube, clear or amber glass reservoir bottle and delivery tip, rubber stopper, rubber bulb, glass air tube, and glass bead


Products ${productFamilyLength}
Catalog Number Mfr. No. Color Includes Price Quantity  
Catalog Number Mfr. No. Color Includes Price Quantity  
03-843-1B Fisherbrand™
720831
Clear Glass buret with central filling tube, clear glass reservoir bottle and delivery tip, rubber stopper, rubber bulb, glass air tube, and glass bead, pinch valve Each for $198.07
03-843-2B Fisherbrand™
720834
Amber Glass buret with central filling tube, amber glass reservoir bottle and delivery tip, rubber stopper, rubber bulb, glass air tube, and glass bead, pinch valve Each for $199.50
Specifications

Description

  • Insert bead into rubber tubing
  • Pinch to stop the flow of liquids

Description & Specifications

Specifications

25mL
Automatic Buret
0.1mL
Glass