Please login to your online account to display your discounted pricing

Fisherbrand™ Automatic Burets for Petroleum Neutralization

Class A, Borosilicate glass construction

$1,603.27 - $1,916.10

Specifications

Certifications/Compliance ASTM D974
Color Clear
Product Type Automatic Buret
For Use With (Application) Petroleum neutralization
Includes 2mm PTFE fluorocarbon resin stopcock, reagent bottle and aspirator bulb
View More Specs
Includes:  PTFE fluorocarbon resin stopcock, reagent bottle and aspirator bulb.
Products
Catalog Number Mfr. No. Standard Taper Joint Capacity (Metric) Subdivision Tolerance Price Quantity    

03-845AAT

 
fisher scientific
TG1829102
24/40 5mL 0.02mL ±0.01mL Each for $1,603.27

03-845AT

 
fisher scientific
TG1829103
24/40 10mL 0.05mL ±0.02mL Each for $1,631.10

03-845BBT

 
fisher scientific
TG1829104
24/40 25mL 0.10mL ±0.03mL Each for $1,636.33

03-845BT

 
fisher scientific
TG1829105
29/42 50mL 0.10mL ±0.05mL Each for $1,916.10
Description & Specifications

Specifications

Certifications/Compliance ASTM D974
Color Clear
Product Type Automatic Buret
For Use With (Application) Petroleum neutralization
Includes 2mm PTFE fluorocarbon resin stopcock, reagent bottle and aspirator bulb
Material Borosilicate Glass

  • Petroleum sample is dissolved in beaker, then titrated with standard acid or base reagent from buret until neutralized
  • Buret reading indicates amount of reagent dispensed to reach neutralization
  • Air openings on buret have drying tubes filled with activated alumina for absorption of vapors and moisture
  • Absorbent can be regenerated by heating

Meet ASTM™ D 974