Please login to your online account to display your discounted pricing

Fisherbrand™ Automatic Burets for Petroleum Neutralization


Class A, Borosilicate glass construction

Manufacturer: fisher scientific  TG1829104

 View more versions of this product

Catalog No. 03-845BBT

  • / Each
 

Description & Specifications

Specifications

Includes 2mm PTFE fluorocarbon resin stopcock, reagent bottle and aspirator bulb
Material Borosilicate Glass
Capacity (Metric) 25mL
Certifications/Compliance ASTM D974
Color Clear
For Use With (Application) Petroleum neutralization
Standard Taper Joint 24/40
Product Type Automatic Buret
Subdivision 0.10mL
Tolerance ±0.03mL