missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Autoclavable Pipet Basins

Eliminate the need for multiple reagent containers—save bench space.

Manufacturer:  Fisherbrand™ 378803001

 View more versions of this product

Catalog No. 13-681-801



Description

Description

Eliminate the need for multiple reagent containers—save bench space.

V-shaped basins facilitate easy withdrawal of sample solutions. Numbered chambers ensure accurate sample reference. Chemical-resistant polypropylene.

12-Channel Pipet Basins

  • Designed for serial dilutions or pipetting individual reagents using up to a 12-channel pipetter
  • Each chamber holds up to 5mL
  • Withstand temperatures from -144° to +218°F (-80° to +121°C)
  • Tight-fitting lid minimizes evaporation or contamination during incubation or storage
  • Sterile or nonsterile; individually wrapped in easy-open bags
  • Dimensions: 5L × 2.25W × 1.2 in.H (12.7 × 5.7 × 3.0cm)
Multichannel Pipet Basin
  • Unique design features three separate compartments: a 50mL single compartment with graduation marks at 10, 25, 40, and 50mL on one side; on the other side, an 8-channel compartment with eight 5mL reagent channels, and a 12-channel compartment with twelve 5mL reagent channels 5mL channels are numbered for easy reagent identification
  • Use with all brands of multichannel pipetters
  • Also ideal for filling multitube blocks and plates
  • Dimensions: 4.75L × 4.3W × 1.1 in.H (12.1 × 10.9 × 2.8cm)

Specifications

Specifications

Yes
5 x 2.25 x 1.2 in. (12.7 x 5.71 x 3.04cm)
3.04 cm
5 in.
Bag
5.71 cm
Non-sterile
12
1.2 in.
Lid
12.7 cm
2.25 in.
Polypropylene
Pipette Basin
Documents