Please login to your online account to display your discounted pricing

Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water


Accuracy meets or exceeds ASTM™ E 100 specifications

Manufacturer: fisherbrand™  116034EC

 View more versions of this product

Catalog No. 11-603-4EC

  • / Each
 

Description & Specifications

Specifications

Subdivisions 0.0005
Increments 0.0005
Temperature of Standardization 60°F
For Use With (Application) Liquids heavier than Water
Specific Gravity Range 0.85 to 0.9
Volume (Metric) 275mL
ASTM Number 86H
Certifications/Compliance 3-point Certification, ASTM E126
Length (Metric) 330mm