Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water
Accuracy meets or exceeds ASTM™ E 100 specifications
Manufacturer:  Fisherbrand™ 116034DC
 View more versions of this product
Catalog No. 11-603-4DC
Specifications
Description
Hydrometer, Specific gravity; Fisherbrand; With ASTM; For liquids lighter than water; 275mL liquid-volume; ASTM No.: 85H; Sp. Gr. range: 0.800 to 0.850; Divison: 0.0005; Approx. length: 330mm; W/3-pt certification
  • Use Cylinder 08-530K
  • Temperature of standardization: 60°F
Description & Specifications
Specifications
0.0005
60°F
Liquids heavier than Water
3-point Certification, ASTM E126
85H
0.0005
0.8 to 0.85
275mL
330mm