Please login to your online account to display your discounted pricing

Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water


Accuracy meets or exceeds ASTM™ E 100 specifications

Manufacturer: fisherbrand™  A1170

 View more versions of this product

Catalog No. 11-603-4C

  • / Each
 

Description & Specifications

Specifications

Subdivisions 0.0005
Increments 0.0005
For Use With (Application) Liquids heavier than Water
Volume (Metric) 275mL
ASTM Number 84H
Length (Metric) 330mm
Specific Gravity Range 0.75 to 0.8
Temperature of Standardization 60°F
Certifications/Compliance ASTM E126
Accessories

Accessories for Catalog Number: 116034C

Item Mfr. No. Item Description Price Per Unit Quantity