Please login to your online account to display your discounted pricing

Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water


Accuracy meets or exceeds ASTM™ E 100 specifications

Manufacturer: fisherbrand™  A1169

 View more versions of this product

Catalog No. 11-603-4B

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Specifications

Increments 0.0005
Subdivisions 0.0005
Certifications/Compliance ASTM E126
ASTM Number 83H
For Use With (Application) Liquids heavier than Water
Length (Metric) 330mm
Temperature of Standardization 60°F
Specific Gravity Range 0.7 to 0.75
Volume (Metric) 275mL

  • Use Cylinder 08-530K
  • Temperature of standardization: 60°F