Please login to your online account to display your discounted pricing

Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water


Accuracy meets or exceeds ASTM™ E 100 specifications

Manufacturer: fisherbrand™  A1168

 View more versions of this product

Catalog No. 11-603-4A

  • / Each
 

Description & Specifications

Specifications

Subdivisions 0.0005
Increments 0.0005
For Use With (Application) Liquids heavier than Water
Temperature of Standardization 60°F
ASTM Number 82H
Certifications/Compliance ASTM E126
Specific Gravity Range 0.65 to 0.7
Volume (Metric) 275mL
Length (Metric) 330mm
Accessories

Accessories for Catalog Number: 116034A

Item Mfr. No. Item Description Price Per Unit Quantity