Please login to your online account to display your discounted pricing

Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Heavier than Water, 260mL Liquid Volume


Accuracy meets or exceeds ASTM E 100 specifications

Manufacturer: fisherbrand™  116038NC

 View more versions of this product

Catalog No. 11-603-8NC

  • / Each
 

Description & Specifications

Specifications

Temperature of Standardization 60°F
Specific Gravity Range 1.6 to 1.65
Volume (Metric) 260mL
ASTM Number 137H
For Use With (Application) Liquids heavier than Water
Increments 0.001
Length (Metric) 260mm
Certifications/Compliance 3-point Certification, ASTM E126