Fisherbrand™ Anti-Static Disposable Medical Blue SMS Lab Coats
Single use SMS lab coats prevent static discharge, static jump, and static decay
$56.67 - $58.36
Specifications
Disposable Yes
Latex Free Yes
Material SMS (Spunbound Melt-Blown Synthetic)
Color Medical blue
No. of Pockets 3
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Length (Metric) Width (Metric) Chest Size Price Quantity  
Catalog Number Mfr. No. Length (Metric) Width (Metric) Chest Size Price Quantity  
17-100-934 Fisherbrand™
17100934
95cm 64.5cm Medium Pack of 10 Each for $56.67
Case of 3 Pack for $170.01
17-100-935 Fisherbrand™
17100935
100cm 66.5cm Large Pack of 10 Each for $56.67
Case of 3 Pack for $170.01
17-100-936 Fisherbrand™
17100936
105cm 70.5cm X-Large Pack of 10 Each for $58.36
Case of 3 Pack for $175.08
17-100-937 Fisherbrand™
17100937
110cm 75cm 2X-Large Pack of 10 Each for $58.36
Case of 3 Pack for $175.08
17-100-939 Fisherbrand™
17100939
125cm 80cm 5X-Large Pack of 10 Each for $58.36
Case of 3 Pack for $175.08
17-100-938 Fisherbrand™
17100938
115cm 78cm 3X-Large Pack of 10 Each for $58.36
Case of 3 Pack for $175.08
17-100-933 Fisherbrand™
17100933
90cm 61.5cm Small Pack of 10 Each for $56.67
Case of 3 Pack for $170.01
Specifications
Description
  • 40gsm SMS anti-static material
  • Knit cuffs and collar
  • Three pockets
  • Single Use
  • Snap closure
  • Blue
Description & Specifications
Specifications
Yes
SMS (Spunbound Melt-Blown Synthetic)
3
Knit
Anti-static lab coat
Yes
Medical blue
Snap
Knit