missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Amber Straight Sided Round Bottle with White Polypropylene PE Foam/PTFE Cap

Ideal for UV sensitive contents. Fisherbrand™ Amber Straight Sided Round Bottle with White Polypropylene PE Foam/PTFE Cap are for general use.

Manufacturer:  Fisherbrand™ 264039

Catalog No. 02-912-045


 

Specifications

Description

Jar, Straight-sided; Fisherbrand; Type III soda-lime glass; Wide mouth; With PTFE-faced PE-lined cap; Screw cap size: 58mm-400; Capacity: 4 oz. (125mL)

  • Straight sided walls allow for the complete removal of contents
  • Not autoclavable
  • Ideal for soil sampling and environmental applications
Description & Specifications

Specifications

4 oz., 125mL
4 oz.
PTFE
Jar and cap
24/PK.
White
Wide
125mL
PTFE
Amber
Soda Lime Glass
Yes
58-400
screw cap