Please login to your online account to display your discounted pricing
Fisherbrand™ Amber Straight-Sided Jars
For light-sensitive soil samples and environmental applications
$63.60 - $90.13
Specifications
Closure Screw
Color Amber
Material Soda Lime Glass
Mouth Wide
View More Specs
Products
Catalog Number Mfr. No. Capacity Includes Closure Size Price Quantity    
02-912-298
fisherbrand™
W014227
4 oz., 125mL Only jar 70-400 Case of 24 Each for $63.60
02-911-859
fisherbrand™
W014225
8 oz., 250mL Jar and cap 70-400 Case of 12 Each for $90.13
Description & Specifications
Specifications
Closure Screw
Color Amber
Material Soda Lime Glass
Mouth Wide
  • Type III soda-lime glass
  • Straight sides allow for complete removal of contents
  • Wide mouth for easy filling
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.