Please login to your online account to display your discounted pricing

Fisherbrand™ Amber Boston Round Bottles with Black Phenolic Rubber Lined Caps


For long term storage. Fisherbrand™ Amber Boston Round Bottles with Black Phenolic Rubber Lined Caps are ideal for use when content integrity and UV protection is important.

Manufacturer: fisherbrand™  263918

 View more versions of this product

Catalog No. 02-912-341

  • / Case of 24 Each
 

Description & Specifications

Specifications

Closure Type Screw
Neck Finish 22-400
Mouth Narrow
Liner Material Rubber
Shape Boston Round
Closure Size 22-400
Capacity (Metric) 120mL
Closure Material Phenolic
Material Glass
Quantity 24/Cs.
Capacity (English) 4 oz.
Color Amber
Capacity 4 oz., 125mL
Closure Liner Rubber