Please login to your online account to display your discounted pricing
Fisherbrand™ Amber Boston Round Bottles with Black Phenolic Rubber Lined Caps
For long term storage. Fisherbrand™ Amber Boston Round Bottles with Black Phenolic Rubber Lined Caps are ideal for use when content integrity and UV protection is important.
$45.17 - $74.70
Specifications
Closure Liner Rubber
Closure Material Phenolic
Closure Type Screw
Color Amber
Material Glass
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Price Quantity    
02-912-343
fisherbrand™
263870
480mL Case of 12 Each for $49.63
02-912-341
fisherbrand™
263918
120mL Case of 24 Each for $57.80
02-912-342
fisherbrand™
263931
240mL Case of 12 Each for $45.17
02-912-344
fisherbrand™
263902
960mL Case of 12 Each for $74.70
Description & Specifications
Specifications
Closure Liner Rubber
Closure Material Phenolic
Closure Type Screw
Color Amber
Material Glass
  • Boston round bottles features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber glass bottles absorb all wavelengths in the ultraviolet spectrum
Black Rubber Lined Phenolic Caps
  • Closure provide consistent adherence to close dimensional tolerance
  • Autoclavable
  • Liner is compliant with FDA 21 CFR 177.2600 Rubber Articles Intended for Repeated Use
  • Liner exhibit good resiliency and resistance to moisture vapor
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.