Fisherbrand™ Amber Boston Round Bottles with White Polypropylene PE Foam/PTFE Lined Caps
For light sensitive solvents, chemicals, and samples. Fisherbrand™ Amber Boston Round Bottles with White Polypropylene PE Foam/PTFE Lined Caps are general use bottles for long term storage of UV sensitive contents.
$31.87 - $69.63
Specifications
Closure Liner PTFE/Polyethylene
Closure Type Screw
Color Amber
Material Glass
Closure Material Polypropylene
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Neck Finish Quantity Price Quantity  
Catalog Number Mfr. No. Capacity Neck Finish Quantity Price Quantity  
02-912-338 Fisherbrand™
263885
1 oz., 30mL 20-400 48/Cs. Case of 48 Each for $63.07
02-911-737 Fisherbrand™
263892
2 oz., 60mL 20-400 24/Cs. Case of 24 Each for $37.53
02-911-739 Fisherbrand™
263903
32 oz., 1000mL 33-400 12/Cs. Case of 12 Each for $69.63
02-911-738 Fisherbrand™
263871
16 oz., 500mL 28-400 12/Cs. Case of 12 Each for $42.17
02-911-740 Fisherbrand™
263919
4 oz., 125mL 22-400 24/Cs. Case of 24 Each for $50.00
02-911-741 Fisherbrand™
263932
8 oz., 250mL 24-400 12/Cs. Case of 12 Each for $31.87
Specifications
Description
Amber Boston Round Bottles
  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Absorbs all wavelengths in the ultraviolet spectrum
Polypropylene PE Foam/PTFE Lined Caps
  • Good impact strength, thermal resistance, and pliability
  • Autoclavable
  • Liners consist of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • Liners are very resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference
Description & Specifications
Specifications
PTFE/Polyethylene
Amber
Polypropylene
Screw
Glass