Fisherbrand™ Amber Boston Round Bottles with White Polypropylene Pulp/Vinyl Lined Caps
Amber bottles are general use bottles ideal for when content integrity and UV protection is important. They ensure integrity for long term storage and offer excellent protection for UV-sensitive contents.
$582.35 - $582.35
Specifications
Closure Liner Poly/Vinyl
Closure Material PTFE
Closure Type screw cap
Closure Size 20-400
Color Amber
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Quantity Price Quantity  
Catalog Number Mfr. No. Capacity Quantity Price Quantity  
02-911-742 Fisherbrand™
W014039
2 oz., 60mL 288/PK. Case for $582.35
Specifications
Description
  • Boston round bottles features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber glass bottles absorb all wavelengths in the ultraviolet spectrum
White Polypropylene Pulp/Vinyl Lined Caps
  • Polypropylene Caps are known for good impact strength, durability, cost effectiveness, thermal resistance, and pliability
  • Caps are autoclavable
  • Liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
Description & Specifications
Specifications
Poly/Vinyl
screw cap
Amber
PTFE
20-400
Glass