Alkacid™ Wide-Range Test Ribbons
Get instant pH values with these handy pocket tester rolls
Manufacturer: Fisher Chemical A979
Description
- Provide five distinct color changes in pH range 2 through 10; color changes: red (pH 2), orange (pH 4), yellow (pH 6), green (pH 8), blue (pH 10)
- Intermediate pH values can be estimated
Order Info
Transparent plastic dispenser with color chart for comparison.
Specifications
Provide wide-range of five distinct color changes in pH range 2 through 10, color changes: red (pH 2), orange (pH 4), yellow (pH 6), green (pH 8), blue (pH 10), intermediate pH values can be estimated, get instant pH values | |
Ribbon | |
2 to 10 |
3 Pack | |
Alkacid™ pH Indicator Paper |