missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Acid-Dispensing Burets

Class A

$1,707.17 - $1,857.23

Specifications

Color Clear
Type Acid Automatic Burette
For Use With (Equipment) Reservoir Bottle
Includes 2 mm PTFE Stopcock, Aspirator Bulb, Reservoir
Material Borosilicate Glass
View More Specs

Includes: PTFE stopcock, reservoir, and aspirator bulb.

Products 2
Catalog Number Mfr. No. Capacity (Metric) Graduations Tolerance Standard Taper Joint Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Graduations Tolerance Standard Taper Joint Price Quantity  
03-846BBT
View Documents
Fisherbrand™
TG1830124
25 mL 0.1 mL ±0.03 mL 24/40
Each for $1,707.17
 
03-846BT
View Documents
Fisherbrand™
TG1830125
50 mL 0.1 mL ±0.05 mL 29/42
Each for $1,857.23
 
Description

Description

  • Precision-grade buret has standard taper joint that fits into neck of reservoir bottle

Specifications

Specifications

Clear
Reservoir Bottle
Borosilicate Glass
Acid Automatic Burette
2 mm PTFE Stopcock, Aspirator Bulb, Reservoir
SDS
Product Certifications