missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Absorption Rectangular Quartz Flow Cells

Choose cells with quartz windows for transformation from the far ultraviolet through infrared

Manufacturer:  Fisherbrand™ 14958157

 View more versions of this product

Catalog No. 14-958-157




Description

Description

  • Compact size
  • All cells are 12.5mm square
  • Fused-on inlet and outlet tubes
  • With two clear windows and two black windows
  • Wavelength range for all cells is 190 to 2600nm and 2850 to 3600nm

Specifications

Specifications

35mm
3.5mm
390µL
Compact Flow Cell
190 to 2600nm and 2850 to 3600nm
12.5mm
10 mm
Documents
Promotions

Promotions