missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Absorption Quartz Semi Micro Rectangular Cells with PTFE Lid, w/PTFE lid

Fit standard spectrophotometer cell holders

Manufacturer:  Fisherbrand™ 14958127

 View more versions of this product

Catalog No. 14-958-127




Description

Description

Fit standard spectrophotometer cell holders

  • All cells are 12.5mm wide
  • 10mm path length rectangular cells
  • Reduce reagent use with Semimicro cells
  • Quartz windows for transmission from far ultraviolet through the infrared

Specifications

Specifications

PTFE
For transmission from far ultraviolet through the infrared
45mm
Rectangular
4mm
1000µL
10 mm
Bottom thickness: 9mm
Standard spectrophotometer cell holder
4/Pk., 8 Pk./Cs.
200 to 2500nm
12.5mm
Quartz
Semi-micro Absorption Cell
Documents
Promotions

Promotions