missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Absorption Cylindrical Cells

Fit standard spectrophotometer cell holders

Manufacturer:  Fisherbrand™ 14958144

 View more versions of this product

Catalog No. 14-958-144




Includes: A filler neck and tapered PTFE stopper (two necks and stoppers on 50 and 100mm lightpath cells).

Description

Description

Fit standard spectrophotometer cell holders

  • All cells are 22mm wide
  • Available size is macro
  • 10, 20, 50 or 100mm path length rectangular cells
  • Available with a variety of window material for wide range of assays
  • Available in two materials e.g. Quartz and Special Optical Glass
  • Quartz windows for transmission from far ultraviolet through the infrared
  • Glass windows for routine work in ultraviolet region and visible region

Specifications

Specifications

35mm
320 to 2500nm
22mm
Optical Glass
Cell
Cylindrical
18.5mm
3100µL
10 mm
Documents
Promotions

Promotions