missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Absorption Cylindrical Cells

Fit standard spectrophotometer cell holders

Manufacturer:  Fisherbrand™ 14958140

 View more versions of this product

Catalog No. 14-958-140




Includes: A filler neck and tapered PTFE stopper (two necks and stoppers on 50 and 100mm lightpath cells).

Description

Description

Fit standard spectrophotometer cell holders

  • All cells are 22mm wide
  • Available size is macro
  • 10, 20, 50 or 100mm path length rectangular cells
  • Available with a variety of window material for wide range of assays
  • Available in two materials e.g. Quartz and Special Optical Glass
  • Quartz windows for transmission from far ultraviolet through the infrared
  • Glass windows for routine work in ultraviolet region and visible region

Specifications

Specifications

35mm
190 to 2600nm and 2850 to 3600nm
22mm
Quartz
Cell
Cylindrical
18.5mm
3100µL
10 mm
Documents
Promotions

Promotions