Please login to your online account to display your discounted pricing

Fisherbrand™ 96-Well Polystyrene Plates


Ideal for antibiotic screens, cell-based assays and screening compounds

Manufacturer: fisherbrand™  N235100

 View more versions of this product

Catalog No. 12-565-500

  • / Case of 80 Each
 

Description & Specifications

Specifications

Lid Yes
Sterile No
No. of Wells 96.0
Quantity 80/Cs.
Well Design Round (U)
Closure Yes
Volume (Metric) Well 250μL
For Use With (Application) Ideal for antibiotic screens, cell based assays and screening compounds
Material Polystyrene
Color Clear
Type 96-Well Assay Microplate
Accessories

Accessories for Catalog Number: 12565500

Item Mfr. No. Item Description Price Per Unit Quantity  
12-565-511
Fisherbrand™ Lid for 96/384 Well Polystyrene Plates
fisherbrand™
N236400
Protect samples from contamination and evaporation during assay processing, incubation or storage Case of 180 Each for $727.29