Please login to your online account to display your discounted pricing

Fisherbrand™ 96-Well Polystyrene Plates


Ideal for antibiotic screens, cell-based assays and screening compounds

Manufacturer: fisherbrand™  N268150

 View more versions of this product

Catalog No. 12-566-622

  • / Case of 180 Each
 

Description & Specifications

Specifications

Lid Yes
Sterile No
No. of Wells 96.0
Closure Yes
Material Polystyrene
Volume (Metric) Well 250μL
Color Clear
For Use With (Application) Ideal for antibiotic screens, cell based assays and screening compounds
Quantity 180/Cs.
Type 96-Well Assay Microplate
Well Design Round (U)