Please login to your online account to display your discounted pricing

Fisherbrand™ 96-Well Polystyrene Plates


Ideal for antibiotic screens, cell-based assays and screening compounds

Manufacturer: fisherbrand™  N237115

 View more versions of this product

Catalog No. 12-566-620

  • / Case of 180 Each
 

Description & Specifications

Specifications

Lid Yes
Sterile No
No. of Wells 96.0
Total Count 96.0
For Use With (Application) Ideal for antibiotic screens, cell based assays and screening compounds
Material Polystyrene
Quantity 180/Cs.
Type 96-Well Assay Microplate
Volume (Metric) Well 350μL
Closure Yes
Color Black
Well Design Flat (F)
Well Shape Triangular
Applications Ideal for antibiotic screens, cell based assays and screening compounds
Well Design Flat (F)
Accessories

Accessories for Catalog Number: 12566620

Item Mfr. No. Item Description Price Per Unit Quantity