Please login to your online account to display your discounted pricing

Fisherbrand™ 96-Well Polystyrene Plates


Ideal for antibiotic screens, cell-based assays and screening compounds

Manufacturer: fisherbrand™  N236113

 View more versions of this product

Catalog No. 12-566-619

  • / Case of 180 Each
 

Description & Specifications

Specifications

Lid Yes
Sterile No
No. of Wells 96.0
Quantity 180/Cs.
Well Design Flat (F)
Closure Yes
Material Polystyrene
Type 96-Well Assay Microplate
Color White
For Use With (Application) Ideal for antibiotic screens, cell based assays and screening compounds
Volume (Metric) Well 350μL