Please login to your online account to display your discounted pricing

Fisherbrand™ 96-Well Polypropylene Microplates


Ideal for storage—feature generous 500μL well volumes

Manufacturer: fisherbrand™  N235500

 View more versions of this product

Catalog No. 12-565-504

  • / Case of 80 Each
 

Description & Specifications

Specifications

For Use With (Application) For storage feature generous 500μL well volumes
Sterile No
Color Yellow
Type 96-Well Storage Microplate
No. of Wells 96
Volume (Metric) Well 500μL
Material Polypropylene
Well Design Round