Please login to your online account to display your discounted pricing

Fisherbrand™ 96-Well Polypropylene Microplates


Ideal for storage—feature generous 500μL well volumes

Manufacturer: fisherbrand™  N235300

 View more versions of this product

Catalog No. 12-565-502

  • / Case of 80 Each
 

Description & Specifications

Specifications

Type 96-Well Storage Microplate
Well Design Round
For Use With (Application) For storage feature generous 500μL well volumes
No. of Wells 96
Color Natural
Material Polypropylene
Volume (Metric) Well 500μL
Sterile No