Please login to your online account to display your discounted pricing

Fisherbrand™ 96-Well Polypropylene Microplates


Ideal for storage—feature generous 500μL well volumes

Manufacturer: fisherbrand™  N267333

 View more versions of this product

Catalog No. 12-566-610

  • / Case of 120 Each
 

Description & Specifications

Specifications

No. of Wells 96
Volume (Metric) Well 500μL
For Use With (Application) For storage feature generous 500μL well volumes
Material Polypropylene
Sterile Yes
Type 96-Well Storage Microplate
Well Design Round
Color Natural
Volume (Metric) Well 500000μL
Well Shape Round
Material Polypropylene