Please login to your online account to display your discounted pricing

Fisherbrand™ 96-Well DeepWell™ Polypropylene Microplates


Ideal for storage because of the low-binding characteristics of polypropylene

Manufacturer: fisherbrand™  N278753

 View more versions of this product

Catalog No. 12-566-613

  • / Case of 50 Each
 

Description & Specifications

Specifications

Color Natural
Material Polypropylene
No. of Wells 96
Volume (Metric) Well 2000μL
For Use With (Application) For storage of low-binding characteristics of polypropylene
Type 96-Well Storage Microplate
Sterile No
Well Design Round