missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ 4 oz. Polypropylene Specimen Containers

Have secure screw-on lids

Manufacturer:  Fisherbrand™ 14828321

 View more versions of this product

Catalog No. 14-828-321



Description

Description

Have secure screw-on lids

  • Convenient 4 oz. capacity
  • Marked with 10cc (mL) and 0.5 oz. increments up to 120cc (mL)
  • Available in sterile or nonsterile
  • Nonsterile container bases are packaged in sleeves (25 each per sleeve); lids are bulk packaged
  • Sterile containers are individually wrapped with secure screw-on lids
  • Identification labels are provided with each case

Order Info

Specimen containers and tops do not come preassembled.

Specifications

Specifications

Non-sterile
White
Secure screw-on lids
Identification labels
100cc
Polypropylene
4.0 oz.
Documents