Please login to your online account to display your discounted pricing

Fisherbrand™ 384-Well ShallowWell Polypropylene Microplates


Small volume applications

Manufacturer: fisherbrand™  N267465

 View more versions of this product

Catalog No. 12-566-618

  • / Case of 100 Each
 

Description & Specifications

Specifications

For Use With (Application) Luminescent assays
Material Polypropylene
No. of Wells 384
Sterile No
Type 384-Well Storage Microplate
Certifications/Compliance ANSI
Volume (Metric) Well 120μL
Well Design Round
Color White
Material Polypropylene
Well Shape Round
Total Count 384.0
Volume (Metric) Well 120μL