Please login to your online account to display your discounted pricing

Fisherbrand™ 384-Well ShallowWell Polypropylene Microplates


Small volume applications

Manufacturer: fisherbrand™  N267463

 View more versions of this product

Catalog No. 12-566-616

  • / Case of 100 Each
 

Description & Specifications

Specifications

Certifications/Compliance ANSI
Color White
For Use With (Application) Storage or colorimetric assays
Material Polypropylene
No. of Wells 384
Sterile Yes
Type 384-Well Storage Microplate
Volume (Metric) Well 58μL
Well Design Round