Fisherbrand™ 384-Well Polystyrene Plates
Process four times as many samples as standard 96-well plates
Supplier: Fisherbrand™ 235700
Description
Optimized for robotic instruments, plate readers and liquid handling systems.
- Highly specified polystyrene
- Ideal for low-volume assays such as colorimetric, enzymatic, cloning, replication, and array, as well as storage
- Flat well bottom surface provides greater area for binding
- Working volume: 10–100μL
- 384-well microplates conserve samples and reagents, providing cost savings and greater screening productivity
- Untreated
- Non-sterile
Order Info
Accommodate 12-565-511 lid (sold separately).
Specifications
Standard | |
16 x 24 | |
Polystyrene | |
10 to 100 μL | |
Non-treated | |
For low volume assays such as colorimetric, enzymatic, cloning, replication and array, as well as storage |
Without Lid | |
Clear | |
384 | |
Non-sterile | |
Rounded Square |