Please login to your online account to display your discounted pricing

Fisherbrand™ 384-Well Polystyrene Plates


Process four times as many samples as standard 96-well plates

Manufacturer: fisherbrand™  N262161

 View more versions of this product

Catalog No. 12-566-625

  • / Case of 100 Each
 

Description & Specifications

Specifications

Lid No
Material Polystyrene
Volume (Metric) Working 0.01 to 0.1μL
Closure No
Sterile No
Type 384-Well Plate
Well Design Triangular
Color Clear
For Use With (Application) For low volume assays such as colorimetric, enzymatic, cloning, replication and array, as well as storage
Closure No
Applications For low volume assays such as colorimetric, enzymatic, cloning, replication and array, as well as storage
Well Shape Triangular