Please login to your online account to display your discounted pricing

Fisherbrand™ 384-Well Polystyrene Plates


Process four times as many samples as standard 96-well plates

Manufacturer: fisherbrand™  N262261

 View more versions of this product

Catalog No. 12-566-624

  • / Case of 100 Each
 

Description & Specifications

Specifications

Lid No
Well Design Rounded Square (C)
Material Polystyrene
Type 384-Well Plate
Sterile No
Color Black
Closure No
For Use With (Application) For low volume assays such as colorimetric, enzymatic, cloning, replication and array, as well as storage
Volume (Metric) Working 0.01 to 0.1μL