Please login to your online account to display your discounted pricing

Fisherbrand™ 384-Well Polypropylene Plates


Feature higher well density for economical use of expensive reagents

Manufacturer: fisherbrand™  N235900

 View more versions of this product

Catalog No. 12-565-508

  • / Case of 120 Each
 

Description & Specifications

Specifications

No. of Wells 384
Material Polypropylene
Sterile No
Type 384-Well Storage Microplate
For Use With (Application) For storage of compounds and nucleic acid libraries
Certifications/Compliance ANSI
Volume (Metric) Well 120μL
Color Red