Please login to your online account to display your discounted pricing

Fisherbrand™ 3/8 in. Dot Labels


Manufacturer: fisherbrand™  FISHC26

 View more versions of this product

Catalog No. 11-010-235

  • / Each
 

Description & Specifications

Specifications

Sterile No
Diameter (English) Outside 0.37 in.
Color Red
Diameter (Metric) 9.55mm
Product Type Dot label roll