Please login to your online account to display your discounted pricing

Fisherbrand™ 3/8 in. Dot Labels


Manufacturer: fisherbrand™  FISHC86

 View more versions of this product

Catalog No. 11-010-228

  • / Each
 

Description & Specifications

Specifications

Diameter (English) 0.37 in.
Product Type Dot label roll
Sterile No
Color Brown
Diameter (Metric) 9.55mm