Please login to your online account to display your discounted pricing

Fisherbrand™ 2 in. Industrial-Grade Vinyl Cleanroom Tape

For light industrial use, clean construction and cleanroom support

$18.99 - $19.04

Specifications

Certifications/Compliance FDA, CID
Format Tape
Product Type Industrial grade cleanroom Tape
Quantity Each
Sterile No
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    

19120-3233

 
fisherbrand™
EGVFP2BR
Brown Each for $18.99
Case of 24 Each for $430.80

19120-3234

 
fisherbrand™
EGVFP2DB
Dark Blue Each for $19.04
Case of 24 Each for $431.76

19120-3241

 
fisherbrand™
EGVFP2TR
Transparent Each for $18.99
Case of 24 Each for $430.80

19095108

 
fisherbrand™
2WHEGVFP
White Each for $19.01
Case of 24 Each for $406.08
Description & Specifications

Specifications

Certifications/Compliance FDA, CID
Format Tape
Product Type Industrial grade cleanroom Tape
Quantity Each
Sterile No
For Use With (Application) Aisle marking, labeling, batching, clean-build projects, light industrial use and food processing applications
Grade Industrial
Length (English) 36 yd.
Length (Metric) 33m
Material Vinyl
Width (English) 2 in.

  • Designed for use in areas where ultraclean tape is not required
  • Forms a strong bond with plastics, polymers and stainless steel without leaving adhesive residue when removed
  • Can be written on with cleanroom pen
  • Processed in a controlled environment
  • Wound on a plastic core and sleeve bagged
  • All rolls are 2 in. wide (5.1cm) by 36 yd. long (33m)

Complies with Federal Specifications PPP-T-66E and CID A-A 1689A