Please login to your online account to display your discounted pricing

Fisherbrand™ 1536-Well Polystyrene Plates


Engineered for high throughput screening, allowing end users to screen four times as many samples in one plates

Manufacturer: fisherbrand™  N264762

 View more versions of this product

Catalog No. 12-566-628

  • / Case of 180 Each
 

Description & Specifications

Specifications

Lid No
Bottom Flat (F)
Closure No
Color White
For Use With (Application) For low volume assays such as colorimetric, enzymatic, cloning, replication and array, as well as storage
Material Polystyrene
Sterile No
Surface Coating Non-Treated
Type 1536-Well Microplate
Volume (Metric) Working 1 to 101μL/well