Please login to your online account to display your discounted pricing

Fisherbrand™ 1536-Well Polystyrene Plates


Engineered for high throughput screening, allowing end users to screen four times as many samples in one plates

Manufacturer: fisherbrand™  N264771

 View more versions of this product

Catalog No. 12-566-627

  • / Case of 180 Each
 

Description & Specifications

Specifications

Lid No
Bottom Flat (F)
Type 1536-Well Microplate
Closure No
For Use With (Application) For low volume assays such as colorimetric, enzymatic, cloning, replication and array, as well as storage
Surface Coating Non-Treated
Volume (Metric) Working 1 to 101μL/well
Sterile No
Material Polystyrene
Color Black