Please login to your online account to display your discounted pricing
Fisherbrand™ 1536-Well Polystyrene Plates
Engineered for high throughput screening, allowing end users to screen four times as many samples in one plates
$889.15 - $966.67
Specifications
Lid No
Sterile No
Bottom Flat (F)
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    
12-566-628
fisherbrand™
N264762
White Case of 180 Each for $889.15
12-566-626
fisherbrand™
N264770
Clear Case of 100 Each for $966.67
12-566-627
fisherbrand™
N264771
Black Case of 180 Each for $898.30
Description & Specifications
Specifications
Lid No
Sterile No
Bottom Flat (F)
Optimized for robotic instruments, plate readers and liquid handling systems
  • Highly specified polystyrene
  • Available in black, white and clear
  • Low plate height optimized for stacking efficiency
  • Working volume range: 1–10lμL/well
  • Untreated
  • Non-sterile
Lids sold seperately