Fisherbrand™ 10/30 Ground Joint Thermometers
Contains red organic liquid against white back glass.
Supplier:  Fisherbrand™ B608041520
 View more versions of this product
Catalog No. 13-201-931
Description
Description
  • 10/30 ground joint with Celsius scale
  • Calibrated for specific immersion depths
  • Individually serialized and packaged
Specifications
Specifications
Partial Immersion
12 in.
Glass
150°C
-10°C to +150°C
10/30 Ground joint with Celsius scale
305 mm
Red Organic Liquid
-10°C
1°C
75 mm