Please login to your online account to display your discounted pricing

Fisherbrand™ 0.5 in. Dot Labels


Manufacturer: fisherbrand™  FISHC36

 View more versions of this product

Catalog No. 11-010-219

  • / Pack of 20 Each
 

Description & Specifications

Specifications

Sterile No
Product Type Dot label sheet
Diameter (English) 0.5 in.
Color Yellow
Diameter (Metric) 12.7mm