Please login to your online account to display your discounted pricing

Fisherbrand™ 0.5 in. Dot Labels


Manufacturer: fisherbrand™  FISHC48

 View more versions of this product

Catalog No. 11-010-251

  • / Each
 

Description & Specifications

Specifications

Sterile No
Diameter (English) Outside 0.5 in.
Diameter (Metric) 12.7mm
Color Violet
Product Type Dot label roll