Please login to your online account to display your discounted pricing

FisherBiotech™ Vertical Electrophoresis Systems


Choice of 10 × 10cm, 16 × 14cm, or 20 × 20cm gel size

Manufacturer: fisherbiotech™  P10DS2XM-FIS

 View more versions of this product

Catalog No. FB-VE20-1

  • / Each
 

Description & Specifications

Specifications

Warranty 3 Years
Width (English) Exterior 6.5 in.
Volume (English) Buffer 38.88 oz.
Length (English) Gel 7.87 in.
Width (Metric) Gel 20cm
Dimension (English) Exterior 6.5 x 12.19 x 9.5 in.
Includes Upper buffer chamber with cooling chamber, lower buffer chamber, safety lid (with attached power supply leads), gaskets, clamp assemblies, gel caster, four blank and two notched glass plates, two 15-well and two 20-well, 1.5mm thick combs, eEight, 1.5mm thick side spacers
Length (Metric) Exterior 30.48cm
Length (Metric) Gel 20cm
Width (English) Gel 7.87 in.
Height (English) Exterior 9 in.
Dimensions (L x W x H) Exterior 12 x 6.5 x 9 in. (30.48 x 16.51 x 23cm)
Gel Dimensions 20 x 20cm
Volume (Metric) 1150mL
Width (Metric) Exterior 16.51cm
Volume (Metric) Buffer 1150mL
Height (Metric) Exterior 23cm
Length (English) Exterior 12 in.