Please login to your online account to display your discounted pricing

FisherBiotech™ Ultraviolet Transilluminators

Variable- or fixed-intensity transilluminators with high-intensity 312nm tubes

Manufacturer: fisherbiotech™  FBTIV614AQ

 View all options for this product

Catalog No. FBTIV614AQ


Description & Specifications


Voltage 115V
Hertz 60Hz
Certifications/Compliance CSA
Height (English) 4.25 in.
Product Type UV Transilluminator
Width (English) 19.25 in.
For Use With (Application) Detecting nanogram amounts of DNA
Filtration Area (English) 6 x 14 in.
Height (Metric) 11cm
Length (English) 13 in.
Intensity Variable
Lighting Type UV
No. of Places 6.0
Wattage 15w
Length (Metric) 33cm
Wavelength 312nm
Width (Metric) 48.89cm
Filtration Area (Metric) 15.2 x 35.6cm
Dimensions (L x W x H) 13 x 19.25 x 4.25 in. (33 x 48.89 x 11cm)
Electrical Requirements 115V 60Hz

  • Six 15w tubes produce a total UV intensity of 8000μw/cm2 to detect nanogram amounts of DNA
  • At 312nm, virtually eliminate DNA degradation caused by 254nm UV light
  • Three sizes of filter areas
  • Unique, removable diffusing screens on all models eliminate confusing UV striations for best photographic results
  • Cooling fan prevents heat buildup

Variable-intensity models

  • Continuously adjustable from 100 to 20% intensity
  • Hinged UV-blocking Safety Cover protects user from hazardous UV radiation
  • Optional removable UV-transmitting Plexiglas™ Filter Protector (Mfr. No. FB-TI-100CQ) shields filter glass from scratches or breakage
Economical, fixed-intensity models
  • Similar to variable-intensity models, but without adjustable intensity feature
  • Timers allow preset of UV exposure time for periods of up to 60 minutes

Fisher Service Division offers several types of service agreements for the equipment featured here. For details on these agreements, equipment repairs, and calibration services, see the front of the catalog. For immediate assistance, call 1-800-395-5442.